Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloromethyl-5-nitrosalicylaldehyde 97%

Supplier: AMBEED, INC
Danger

A196292-1G A196292-5G A196292-1G A196292-25G A196292-5G
103816-834EA 19 USD InStock
103816-834 103847-446 77302-512 77302-514 77302-516
3-Chloromethyl-5-nitrosalicylaldehyde 97%
3-Chloromethyl-5-nitrosalicylaldehyde
Precaución: for Research Use Only


Formula: C₈H₆ClNO₄
Boiling Pt: 372.8 °C (760 mm Hg)
Flash Pt: 179.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191331
CAS Number: 16644-30-7
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty