Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A244616-5G A244616-25G A244616-100G A244616-10G A244616-500G A244616-5G
103851-426EA 23 USD InStock
103851-426 77090-636 77276-112 77276-114 77276-116 77304-326
3-(Hydroxymethyl)phenylboronic acid 98%
3-(Hydroxymethyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₉BO₃
MW: 151.96 g/mol
Boiling Pt: 380.5 °C (760 mm Hg)
Melting Pt: 95…99 °C
Flash Pt: 183.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01317846
CAS Number: 87199-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty