Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylsulfonyl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A321658-1G A321658-5G A321658-10G A321658-250MG
103825-286EA 19 USD InStock
103825-286 103836-218 76839-130 76875-856 76875-858
3-(Methylsulfonyl)propanoic acid 98%
3-(Methylsulfonyl)propanoic acid
Precaución: for Research Use Only


Formula: C₄H₈O₄S
Boiling Pt: 409 °C (760 mm Hg)
Melting Pt: 102…105 °C
Flash Pt: 201.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01697360
CAS Number: 645-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty