Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Hydroxyprop-1-yl)piperidine 97%

Supplier: AMBEED, INC
Warning

A303797-1G A303797-5G A303797-1G A303797-25G A303797-5G A303797-100G A303797-10G
103814-392EA 6 USD InStock
103814-392 103835-558 76838-240 76838-242 76838-244 77239-292 77239-294
1-(3-Hydroxyprop-1-yl)piperidine 97%
1-(3-Hydroxyprop-1-yl)piperidine
Precaución: for Research Use Only


Formula: C₈H₁₇NO
Boiling Pt: 94…95 °C (0.5 mm Hg)
Flash Pt: 97.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00023781
CAS Number: 104-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty