Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Piperidin-4-yl)pyridine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A274596-1G A274596-100MG A274596-1G
103813-202EA 693 USD InStock
103813-202 103853-130 77166-542 77305-166
3-(Piperidin-4-yl)pyridine hydrochloride 97%
3-(Piperidin-4-yl)pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD16988818
CAS Number: 690261-73-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty