Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyrrolidinophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A112715-1G A112715-100MG A112715-250MG A112715-1G A112715-5G
103798-082EA 35 USD InStock
103798-082 103823-922 76995-306 76995-308 77310-732 77310-734
3-Pyrrolidinophenylboronic acid 97%
3-Pyrrolidinophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₄BNO₂
MW: 191.035 g/mol
Boiling Pt: 403.2 °C (760 mm Hg)
Flash Pt: 197.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095125
CAS Number: 659731-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty