Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(tert-Butyl)-1H-pyrazole-5-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A313034-1G A313034-5G A313034-100MG A313034-250MG
103814-712EA 62 USD InStock
103814-712 103835-902 76838-722 77002-722 77002-724
3-(tert-Butyl)-1H-pyrazole-5-carboxylic acid 95%
3-(tert-Butyl)-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₂
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00462163
CAS Number: 83405-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty