Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-tert-Butyl-4-methoxyphenol 97%

Supplier: AMBEED, INC
Warning

A186045-1G A186045-5G A186045-100MG A186045-10G A186045-250MG A186045-1G A186045-5G A186045-25G
103815-902EA 19 USD InStock
103815-902 103846-476 77274-290 77274-292 77274-294 77302-008 77302-010 77748-206
3-tert-Butyl-4-methoxyphenol 97%
3-tert-Butyl-4-methoxyphenol
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₂
MW: 180.25
Boiling Pt: 293.3 °C (760 mm Hg)
Melting Pt: 64 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00057667
CAS Number: 88-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty