Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-tert-Butylaniline 98%

Supplier: AMBEED, INC
Warning

A136818-1G A136818-100G A136818-10G A136818-1G A136818-25G A136818-5G
103802-356EA 9 USD InStock
103802-356 76688-830 76688-832 77298-868 77298-870 77315-276
3-tert-Butylaniline 98%
3-tert-Butylaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₅N
Boiling Pt: 236.6 °C (760 mm Hg)
Flash Pt: 100.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00125078
CAS Number: 5369-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty