Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)dibenzo[b,e]oxepin-11(6H)-one 98%

Supplier: AMBEED, INC
Warning

A130702-1G A130702-100MG A130702-250MG
103801-464EA 251 USD InStock
103801-464 76811-158 76811-160
3-(Trifluoromethyl)dibenzo[b,e]oxepin-11(6H)-one 98%
3-(Trifluoromethyl)dibenzo[b,e]oxepin-11(6H)-one
Precaución: for Research Use Only


Formula: C₁₅H₉F₃O₂
Boiling Pt: 394.2±42.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10565665
CAS Number: 4504-94-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty