Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)isonicotinic acid 97%

Supplier: AMBEED, INC
Warning

A167742-1G A167742-5G A167742-1G A167742-250MG
103806-214EA 34 USD InStock
103806-214 103844-474 77081-506 77273-598
3-(Trifluoromethyl)isonicotinic acid 97%
3-(Trifluoromethyl)isonicotinic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234940
CAS Number: 590371-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty