Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Difluorocyclobutanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A310963-1G A310963-1G A310963-25G A310963-5G A310963-100MG A310963-10G A310963-250MG
103814-642EA 9 USD InStock
103814-642 103835-828 76838-602 76838-604 76838-606 77277-522 77277-524 77277-526
3,3-Difluorocyclobutanamine hydrochloride 97%
3,3-Difluorocyclobutanamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₈ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD10001382
CAS Number: 637031-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty