Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Difluoropiperidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A113721-1G A113721-5G A113721-10G A113721-25G A113721-1G A113721-5G
103798-278EA 41 USD InStock
103798-278 103824-120 77271-058 77271-060 77271-062 77310-932 77310-934
3,3-Difluoropiperidine hydrochloride 98%
3,3-Difluoropiperidine hydrochloride
Precaución: for Research Use Only


Formula: C5H10ClF2N
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD03452800
CAS Number: 496807-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty