Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethylazetidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A324906-1G A324906-5G A324906-100MG
103825-386EA 92 USD InStock
103825-386 103836-328 76839-250 77872-463
3,3-Dimethylazetidine hydrochloride 97%
3,3-Dimethylazetidine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClN
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD10565785
CAS Number: 89381-03-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty