Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethylcyclohexanone 97%

Supplier: AMBEED, INC
Danger

A460054-5G A460054-25G A460054-5G A460054-10G A460054-1G
103838-988EA 7 USD InStock
103838-988 76842-866 76842-868 77279-254 77279-256
3,3-Dimethylcyclohexanone 97%
3,3-Dimethylcyclohexanone
Precaución: for Research Use Only


Formula: C₈H₁₄O
Boiling Pt: 170.6 °C (760 mm Hg)
Flash Pt: 59.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00040178
CAS Number: 2979-19-3
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty