Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trimethoxycinnamic acid 98%

Supplier: AMBEED, INC
Warning

A157321-5G A157321-100G A157321-25G A157321-5G
103843-188EA 6 USD InStock
103843-188 76768-402 77300-252 77319-008
3,4,5-Trimethoxycinnamic acid 98%
3,4,5-Trimethoxycinnamic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₅
Boiling Pt: 396.4 °C (760 mm Hg)
Melting Pt: 125…127 °C
Flash Pt: 151.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004388
CAS Number: 90-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty