Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diaminobenzonitrile 97%

Supplier: AMBEED, INC
Warning

A182479-1G A182479-1G A182479-25G A182479-5G A182479-100G A182479-10G
103807-718EA 6 USD InStock
103807-718 103846-090 77083-634 77083-636 77083-638 77138-770 77138-772
3,4-Diaminobenzonitrile 97%
3,4-Diaminobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 368.7 °C (760 mm Hg)
Melting Pt: 144…148 °C
Flash Pt: 176.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00723901
CAS Number: 17626-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty