Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diethoxyaniline 97%

Supplier: AMBEED, INC
Warning

A336580-1G A336580-5G A336580-10G A336580-250MG
103825-726EA 13 USD InStock
103825-726 103836-698 76839-706 76839-708 77277-904 77277-906
3,4-Diethoxyaniline 97%
3,4-Diethoxyaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂
MW: 181.24 g/mol
Boiling Pt: 294.5 °C (760 mm Hg)
Melting Pt: 45…50 °C
Flash Pt: 143.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00026782
CAS Number: 39052-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty