Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A306986-1000G A306986-25G A306986-100G A306986-10G A306986-500G
103810-046EA 206 USD InStock
103810-046 76838-398 76838-400 77141-624 77141-626 77141-628
3,4-Difluorobenzoic acid 97%
3,4-Difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00011672
CAS Number: 455-86-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty