Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-1-pyrrolidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A121758-1G A121758-100G A121758-250MG A121758-25G A121758-1G A121758-5G
103800-130EA 10 USD InStock
103800-130 77108-604 77108-606 77108-608 77312-256 77312-258
2-Amino-1-pyrrolidine hydrochloride 95%
2-Amino-1-pyrrolidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₉ClN₂
Boiling Pt: 211 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00790772
CAS Number: 7544-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty