Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-5-hydroxy-1(2H)-isoquinolinone 95%

Supplier: AMBEED, INC
Warning

A330179-50MG A330179-100MG A330179-250MG
103799-600EA 15 USD InStock
103799-600 103836-504 76978-220 76978-222
3,4-Dihydro-5-hydroxy-1(2H)-isoquinolinone 95%
3,4-Dihydro-5-hydroxy-1(2H)-isoquinolinone
Precaución: for Research Use Only


Formula: C₉H₉NO₂
Boiling Pt: 473.9 °C (760 mm Hg)
Flash Pt: 240.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02258901
CAS Number: 56469-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty