Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Pyridinedicarboximide 97%

Supplier: AMBEED, INC
Warning

A101675-1G A101675-5G A101675-100G A101675-10G A101675-25G A101675-1G A101675-5G
103796-084EA 7 USD InStock
103796-084 103821-780 77270-260 77270-262 77270-264 77308-702 77308-704
3,4-Pyridinedicarboximide 97%
3,4-Pyridinedicarboximide
Precaución: for Research Use Only


Formula: C₇H₄N₂O₂
Melting Pt: 230…232 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013439
CAS Number: 4664-01-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty