Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A660511-100G A660511-10G A660511-500G A660511-25G
103840-446EA 11 USD InStock
103840-446 76697-036 76697-038 76697-040 76844-638
3,5-Bis(trifluoromethyl)benzoic acid 98%
3,5-Bis(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260
Melting Pt: 142…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00000388
CAS Number: 725-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty