Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromobenzonitrile 98%

Supplier: AMBEED, INC
Danger

A225869-1G A225869-5G A225869-10G A225869-25G A225869-5G A225869-1G
103819-208EA 9 USD InStock
103819-208 103850-110 76826-306 77088-828 77088-830 77303-754
3,5-Dibromobenzonitrile 98%
3,5-Dibromobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃Br₂N
Boiling Pt: 261.2 °C (760 mm Hg)
Melting Pt: 98 °C
Flash Pt: 111.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00178770
CAS Number: 97165-77-0
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty