Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromophenol 98%

Supplier: AMBEED, INC
Warning

A463585-5G A463585-100G A463585-10G A463585-250MG A463585-500G A463585-1G A463585-25G A463585-5G
103827-956EA 6 USD InStock
103827-956 103839-032 76818-438 76818-440 76818-442 76818-444 76842-932 76842-934 76842-936
3,5-Dibromophenol 98%
3,5-Dibromophenol
Precaución: for Research Use Only


Formula: C₆H₄Br₂O
Boiling Pt: 278.1 °C (760 mm Hg)
Melting Pt: 79…83 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD06411376
CAS Number: 626-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty