Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorobenzamidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A161817-1G A161817-5G A161817-100MG A161817-250MG A161817-1G A161817-5G
103805-540EA 72 USD InStock
103805-540 103843-782 77011-228 77011-230 77300-556 77300-558
3,5-Difluorobenzamidine hydrochloride 95%
3,5-Difluorobenzamidine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₇ClF₂N₂
Storage Temperature: Ambient
MDL Number: MFCD04114449
CAS Number: 144797-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty