Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A139501-1000G A139501-100G A139501-10G A139501-25G
103808-592EA 437 USD InStock
103808-592 76767-712 76767-714 77299-078
3,5-Difluorobenzoic acid 97%
3,5-Difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄F₂O₂
Boiling Pt: 243.2 °C (760 mm Hg)
Melting Pt: 121…123 °C
Flash Pt: 100.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00010323
CAS Number: 455-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty