Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-DL-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A193474-1G A193474-1G A193474-250MG A193474-5G
103816-542EA 19 USD InStock
103816-542 77085-066 77274-548 77274-550
3,5-Difluoro-DL-phenylalanine 97%
3,5-Difluoro-DL-phenylalanine
Precaución: for Research Use Only


Formula: C₉H₉F₂NO₂
MW: 201.1701 g/mol
Boiling Pt: 295.1 °C (760 mm Hg)
Melting Pt: 269.5…273.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00061308
CAS Number: 32133-37-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty