Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-4-pyridinamine 98%

Supplier: AMBEED, INC
Warning

A158809-1G A158809-5G A158809-10G A158809-250MG A158809-1G A158809-25G
103805-176EA 21 USD InStock
103805-176 103843-390 76997-684 76997-686 77300-370 77319-244
3,5-Difluoro-4-pyridinamine 98%
3,5-Difluoro-4-pyridinamine
Precaución: for Research Use Only


Formula: C₅H₄F₂N₂
Melting Pt: 102…105 °C
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD01862101
CAS Number: 159783-22-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty