Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A160645-1000G A160645-100G A160645-500G A160645-1000G A160645-25G
103808-886EA 55 USD InStock
103808-886 77138-134 77138-136 77319-580 77319-582
3,5-Dimethyl-1H-pyrazole 98%
3,5-Dimethyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₅H₈N₂
Boiling Pt: 218 °C (760 mm Hg)
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005243
CAS Number: 67-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty