Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-1,2,4-triazole 97%

Supplier: AMBEED, INC
Warning

A213712-1G A213712-25G A213712-5G A213712-100G A213712-10G
103818-318EA 11 USD InStock
103818-318 103849-026 77087-538 77087-540 77748-332 77748-333
3,5-Dimethyl-1,2,4-triazole 97%
3,5-Dimethyl-1,2,4-triazole
Precaución: for Research Use Only


Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 255.4 °C (760 mm Hg)
Melting Pt: 143…147 °C
Flash Pt: 123.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00656686
CAS Number: 7343-34-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty