Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylbenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A198986-1G A198986-5G A198986-1G A198986-25G A198986-5G A198986-100G A198986-10G A198986-500G
103817-064EA 9 USD InStock
103817-064 103847-688 77085-714 77085-716 77085-718 77248-820 77248-822 77746-248
3,5-Dimethylbenzaldehyde 98%
3,5-Dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O
Boiling Pt: 221 °C (760 mm Hg)
Melting Pt: 2…8 °C
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082777
CAS Number: 5779-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty