Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A116412-1G A116412-5G A116412-100G A116412-10G
103798-730EA 9 USD InStock
103798-730 103824-596 76687-898 76687-900
3,5-Dimethylbenzonitrile 97%
3,5-Dimethylbenzonitrile
Precaución: for Research Use Only


Formula: C₉H₉N
Boiling Pt: 243.1 °C (760 mm Hg)
Melting Pt: 42…43 °C
Flash Pt: 100.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00060670
CAS Number: 22445-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty