Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dimethylpyridazine 97%

Supplier: AMBEED, INC
Warning

A354810-5G A354810-1G A354810-10G A354810-250MG
103826-132EA 44 USD InStock
103826-132 103837-136 76840-264 76989-714 76989-716
3,6-Dimethylpyridazine 97%
3,6-Dimethylpyridazine
Precaución: for Research Use Only


Formula: C₆H₈N₂
Boiling Pt: 228 °C (760 mm Hg)
Melting Pt: 32…33 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD06637608
CAS Number: 1632-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty