Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,7-dihydro-6H-purine-6-thione 98%

Supplier: AMBEED, INC
Warning

A262086-1G A262086-25G A262086-5G A262086-10G A262086-50MG A262086-1G A262086-100G
103812-554EA 12 USD InStock
103812-554 103852-448 77092-062 77092-064 77098-840 77098-842 77304-846 77685-745
3,7-dihydro-6H-purine-6-thione 98%
3,7-dihydro-6H-purine-6-thione
Precaución: for Research Use Only


Formula: C₅H₄N₄S
Boiling Pt: 471.018 °C (760 mm Hg)
Melting Pt: 241…244 °C
Flash Pt: 250.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233552
CAS Number: 50-44-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty