Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetylindole 97%

Supplier: AMBEED, INC
Warning

A172588-1G A172588-100G A172588-10G A172588-1G A172588-25G A172588-5G
103806-730EA 6 USD InStock
103806-730 103845-018 76769-280 76769-282 77082-216 77082-218 77082-220
3-Acetylindole 97%
3-Acetylindole
Precaución: for Research Use Only


Formula: C₁₀H₉NO
Boiling Pt: 333.9 °C (760 mm Hg)
Melting Pt: 188…192 °C
Flash Pt: 163.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005626
CAS Number: 703-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty