Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-pyrazolecarbonitrile 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A190486-5G A190486-100G A190486-10G A190486-500G A190486-25G A190486-5G A190486-1000G
103846-896EA 6 USD InStock
103846-896 76825-258 76825-260 76825-262 77084-692 77302-224 77683-498
3-Amino-4-pyrazolecarbonitrile 97%, Ambeed.Inc
3-Amino-4-pyrazolecarbonitrile
Precaución: For research use only.


Formula: C₄H₄N₄
Boiling Pt: 476.4 °C (760 mm Hg)
Flash Pt: 241.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005237
CAS Number: 16617-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty