Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-3-Aminoisobutyric acid hydrate 95%

Supplier: AMBEED, INC
Warning

A823876-5G A823876-250MG A823876-1G
103841-150EA 84 USD InStock
103841-150 77006-702 77308-406
DL-3-Aminoisobutyric acid hydrate 95%
DL-3-Aminoisobutyric acid hydrate
Precaución: for Research Use Only


Formula: C₄H₉NO₂·H₂O
MW: 103.12 g/mol
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00008145
CAS Number: 144-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty