Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4,6-dichloropicolinic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A214715-1G A214715-5G A214715-1G A214715-5G A214715-100MG A214715-250MG
103818-428EA 101 USD InStock
103818-428 103849-132 77303-370 77303-372 77683-611 77683-612
3-Amino-4,6-dichloropicolinic acid 95%, Ambeed.Inc
3-Amino-4,6-dichloropicolinic acid
Precaución: For research use only.


Formula: C₆H₄Cl₂N₂O₂
Boiling Pt: 472.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD24499074
CAS Number: 1073182-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty