Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-(4-phenoxyphenyl)pyrazole-4-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A464244-5G A464244-10G A464244-250MG A464244-1G A464244-25G A464244-5G
103827-962EA 11 USD InStock
103827-962 103839-040 76829-866 76829-868 76829-870 76842-940 76842-942 76842-944
3-Amino-5-(4-phenoxyphenyl)pyrazole-4-carbonitrile 97%
3-Amino-5-(4-phenoxyphenyl)pyrazole-4-carbonitrile
Precaución: for Research Use Only


Formula: C₁₆H₁₂N₄O
Boiling Pt: 585.3±50.0 °C (760 mm Hg)
Flash Pt: 307.8 °C
Storage Temperature: Ambient
MDL Number: MFCD20270361
CAS Number: 330792-70-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty