Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromopyridin-2(1H)-one 98%

Supplier: AMBEED, INC
Danger

A178823-1G A178823-5G A178823-10G A178823-250MG A178823-25G A178823-5G
103807-370EA 9 USD InStock
103807-370 103845-710 76974-350 76974-352 77083-152 77083-154
3-Amino-5-bromopyridin-2(1H)-one 98%
3-Amino-5-bromopyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C5H5BrN2O
Boiling Pt: 295.1 °C (760 mm Hg)
Flash Pt: 132.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840438
CAS Number: 98786-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty