Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-tert-butylisoxazole 97%

Supplier: AMBEED, INC
Warning

A112824-1G A112824-5G A112824-10G A112824-25G A112824-1G A112824-5G
103798-114EA 13 USD InStock
103798-114 103823-950 77108-100 77108-102 77310-772 77310-774
3-Amino-5-tert-butylisoxazole 97%
3-Amino-5-tert-butylisoxazole
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O
MW: 140.19
Boiling Pt: 248.1 °C (760 mm Hg)
Melting Pt: 110…114 °C
Flash Pt: 103.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00055620
CAS Number: 55809-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty