Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-tert-butyl-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A105905-100G A105905-25G
103822-714EA 7 USD InStock
103822-714 77107-610 77107-612
3-Amino-5-tert-butyl-1H-pyrazole 97%
3-Amino-5-tert-butyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₇H₁₃N₃
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00067987
CAS Number: 82560-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty