Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-6-bromopyrazine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A257308-1G A257308-5G A257308-1G A257308-25G A257308-5G A257308-10G A257308-250MG
103812-294EA 10 USD InStock
103812-294 103852-162 77091-670 77091-672 77091-674 77276-452 77276-454
3-Amino-6-bromopyrazine-2-carboxylic acid 95%
3-Amino-6-bromopyrazine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₅H₄BrN₃O₂
Boiling Pt: 397.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD08275681
CAS Number: 486424-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty