Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminoisonicotinic acid 97% contains < 5% AcOH

Supplier: AMBEED, INC
Warning

A117914-1G A117914-5G A117914-100G A117914-10G A117914-250MG A117914-5G
103798-952EA 12 USD InStock
103798-952 103824-842 77271-318 77271-320 77271-322 77311-622
3-Aminoisonicotinic acid 97% contains < 5% AcOH
3-Aminoisonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₆N₂O₂
Melting Pt: 305…310 °C
Flash Pt: 254.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00137840
CAS Number: 7579-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty