Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminophenylboronic acid monohydrate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A186407-5G A186407-10G A186407-25G A186407-5G A186407-100G
103815-938EA 12 USD InStock
103815-938 103846-520 77012-142 77084-180 77084-182 77683-477
3-Aminophenylboronic acid monohydrate 98%, Ambeed.Inc
3-Aminophenylboronic acid monohydrate
Precaución: For research use only.


Formula: C₆H₈BNO₂·1H₂O
MW: 154.96 g/mol
Melting Pt: 93…96 °C
Flash Pt: 181.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00149554
CAS Number: 206658-89-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty