Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Boc-Amino-2,6-dioxopiperidine 98%

Supplier: AMBEED, INC
Warning

A235536-1G A235536-1G A235536-5G A235536-100G A235536-10G A235536-250MG
103811-020EA 14 USD InStock
103811-020 103850-788 77089-798 77089-800 77275-874 77275-876 77275-878
3-Boc-Amino-2,6-dioxopiperidine 98%
3-Boc-Amino-2,6-dioxopiperidine
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₄
Boiling Pt: 423 °C (760 mm Hg)
Flash Pt: 209.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275101
CAS Number: 31140-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty