Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Carboxy-2-fluorophenylboronic acid 96%

Supplier: AMBEED, INC
Warning

A152437-1G A152437-5G A152437-250MG A152437-5G A152437-1G
103804-392EA 31 USD InStock
103804-392 103842-560 77028-210 77299-950 77318-174
5-Carboxy-2-fluorophenylboronic acid 96%
5-Carboxy-2-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BFO₄
MW: 183.9300 g/mol
Boiling Pt: 432.896 °C (760 mm Hg)
Flash Pt: 215.608 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436065
CAS Number: 874219-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty