Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1,5-naphthyridine 97%

Supplier: AMBEED, INC
Warning

A380695-5G A380695-1G A380695-100MG A380695-250MG
103826-676EA 110 USD InStock
103826-676 103837-710 76841-032 77278-532 77278-534
3-Bromo-1,5-naphthyridine 97%
3-Bromo-1,5-naphthyridine
Precaución: for Research Use Only


Formula: C8H5BrN2
MW: 209.0400 g/mol
Boiling Pt: 289 °C (760 mm Hg)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD11858569
CAS Number: 17965-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty