Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1-methyl-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A100255-1G A100255-5G A100255-10G A100255-25G A100255-1G A100255-5G
103795-324EA 9 USD InStock
103795-324 103821-034 77233-030 77233-032 77282-464 77282-466
3-Bromo-1-methyl-1H-pyrazole 98%
3-Bromo-1-methyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C4H5BrN2
MW: 161.0000 g/mol
Boiling Pt: 197 °C (760 mm Hg)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD11110668
CAS Number: 151049-87-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty